1-Benzofuran-2-carbonitrile 95%

Supplier: Ambeed
Warning

A242547-1G A242547-25G A242547-5G A242547-10G A242547-250MG
103811-474EA 47 USD
103811-474 103851-288 77090-466 77090-468 77090-470 77261-824 77261-826
1-Benzofuran-2-carbonitrile 95%
1-Benzofuran-2-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₅NO
MW: 143.15 g/mol
Boiling Pt: 260.8 °C (760 mm Hg)
Flash Pt: 111.556 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01659779
CAS Number: 41717-32-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR