3-(Dimethylamino)-1-(2-furyl)prop-2-en-1-one 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Dimethylamino)-1-(furan-2-yl)prop-2-en-1-one

A124739-1G
77258-220EA 16.48 USD
77258-220
3-(Dimethylamino)-1-(2-furyl)prop-2-en-1-one 97%
3-(Dimethylamino)-1-(2-furyl)prop-2-en-1-one

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.189 g/mol
MDL Number: MFCD00121186
CAS Number: 17168-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR