(R)-3-Methyl-1-phenylbutan-1-amine 97%

Supplier: Ambeed
Warning

A102129-250MG
77257-274EA 91 USD
77257-274
(R)-3-Methyl-1-phenylbutan-1-amine 97%
(R)-3-Methyl-1-phenylbutan-1-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₇N
MW: 163.25 g/mol
MDL Number: MFCD06762329
CAS Number: 1173110-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR