2,3-Dihydro-1-benzofuran-2-ylmethanol 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydro-2-(hydroxymethyl)benzo[b]furan, (2,3-Dihydrobenzofuran-2-yl)methanol

A267964-250MG
103812-888EA 94 USD
103812-888 103852-788 77250-280
2,3-Dihydro-1-benzofuran-2-ylmethanol 98%
2,3-Dihydro-1-benzofuran-2-ylmethanol

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 272 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03086171
CAS Number: 66158-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR