Ethyl-4-methyl-4-pentenoate 96%

Supplier: Ambeed
Danger

Synonyms: Ethyl 4-methylpent-4-enoate

A560973-100MG A560973-1G A560973-250MG A560973-5G
77242-056EA 9 USD
77242-056 77242-058 77242-060 77569-082
Ethyl-4-methyl-4-pentenoate 96%
Ethyl-4-methyl-4-pentenoate

Precaución: For research use only.

Formula: C₈H₁₄O₂
MW: 142.198 g/mol
MDL Number: MFCD00075160
CAS Number: 4911-54-0
UN: 3272
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR