(4-Cyclopropyl-1,3-thiazol-2-yl)acetonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Cyclopropylthiazol-2-yl)acetonitrile

A304404-100MG
77239-308EA 48 USD
77239-308
(4-Cyclopropyl-1,3-thiazol-2-yl)acetonitrile 97%
(4-Cyclopropyl-1,3-thiazol-2-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂S
MW: 164.23 g/mol
MDL Number: MFCD19596754
CAS Number: 1028352-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR