Emodin 98%

Supplier: Ambeed
Warning

Synonyms: 1,3,8-Trihydroxy-6-methylanthracene-9,10-dione, 1,3,8-Trihydroxy-6-methylanthraquinone

A178654-100G A178654-10G A178654-1G A178654-1MG A178654-250MG A178654-50MG A178654-25G A178654-5G
103845-700EA 32 USD
103845-700 77236-452 77236-454 77236-456 77236-458 77236-460 77236-462 77301-570 77301-572
Emodin 98%
Emodin

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₅
MW: 270.24 g/mol
Melting Pt: 253 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001207
CAS Number: 518-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR