GlyH-101 98+%

Supplier: Ambeed
Warning

Synonyms: CFTR Inhibitor, N'-(3,5-Dibromo-2,4-dihydroxybenzylidene)-2-(naphthalen-2-ylamino)acetohydrazide, Cystic Fibrosis Transmembrane Conductance Regulator Inhibitor II, 2-​[(3,​5-​dibromo-​2,​4-​dihydroxyphenyl)methylene]hydrazide N-​2-​naphthalenyl-​glycine

A128948-10MG A128948-1MG A128948-2MG A128948-50MG A128948-5MG
77234-384EA 32 USD
77234-384 77234-386 77234-388 77234-390 77234-392
GlyH-101 98+%
GlyH-101

Precaución: for Research Use Only

Formula: C₁₉H₁₅Br₂N₃O₃
MW: 493.15 g/mol
MDL Number: MFCD01532954
CAS Number: 328541-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR