Staticide® ACL Polyester Knit Wipes
Supplier: ACL Staticide
8399PK
77200-700CS 390.41 USD
77200-700
Staticide® ACL Polyester Knit Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Polyester knit wipers are are an economical choice for those seeking an all-purpose, lint-free wipe with no fiber loss.
  • 100% pure polyester
  • Basis weight: 120 g/m²
  • Free of silicon, amides, and DOP contamination
  • Laser-sealed edges
  • Class 100 laundered and packaged
  • ISO Class 5 - 7 (Class 100 - 10,000) environments
Great for critical environments and process cleaning, these polyester knit wipes are lightweight and super-soft. The material will not scratch surfaces and offers ultra-low particle and fiber generation.