Staticide® 8101 Economy Wrist Strap Cord, 4 mm to Banana
Supplier: ACL Staticide
8101
77200-578EA 40.54 USD
77200-578
Staticide® 8101 Economy Wrist Strap Cord, 4 mm to Banana
Grounders Grounding Wires
Economical grounding cord with 1-meg ohm resistor.
  • 6 foot cord with flexible strain relief assembly to prevent breakage
  • Recommended for use with 4 mm wrist strap
  • 4 mm socket-to-banana
  • Includes alligator clip
  • Anti-static nylon
The 8101 economy wrist strap cord is designed for use with single-wire wrist straps with 4 mm male stud.
Envase: 6 cords per pack.