1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium tetrafluoroborate 98,5%

Supplier: Ambeed
Danger

Synonyms: Chloro-N,N,N',N'-bis(tetramethylene)formamidinium Tetrafluoroborate, TPyICU, 1-(Chloro(pyrrolidin-1-yl)methylene)pyrrolidin-1-ium tetrafluoroborate

A892420-1G
77200-182EA 19 USD
77200-182
1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium tetrafluoroborate 98,5%
1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₉H₁₆BClF₄N₂
MW: 274.49 g/mol
MDL Number: MFCD00191334
CAS Number: 115007-14-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR