2-(3-Fluoropyridin-2-yl)-2-methylpropanoic acid 97%

Supplier: Ambeed
Warning

A561142-250MG
77197-620EA 121.54 USD
77197-620
2-(3-Fluoropyridin-2-yl)-2-methylpropanoic acid 97%
2-(3-Fluoropyridin-2-yl)-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MW: 183.17 g/mol
MDL Number: MFCD26096781
CAS Number: 1402232-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR