O-Phenetidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-Ethoxyaniline Hydrochloride

A180184-1G
77191-662EA 14 USD
77191-662
O-Phenetidine hydrochloride 97%
O-Phenetidine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 173.64 g/mol
MDL Number: MFCD00060217
CAS Number: 89808-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR