LY-411575 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((S)-2-(3,5-Difluorophenyl)-2-hydroxyacetamido)-N-((S)-5-methyl-6-oxo-6,7-dihydro-5H-dibenzo[b,d]azepin-7-yl)propanamide

A140795-10MG A140795-1MG A140795-25MG A140795-5MG
77190-400EA 95 USD
77190-400 77190-402 77190-404 77190-406
LY-411575 98%
LY-411575

Precaución: for Research Use Only

Formula: C₂₆H₂₃F₂N₃O₄
MW: 479.48 g/mol
MDL Number: MFCD09832551
CAS Number: 209984-57-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR