N,N'-Di-Boc-guanidine 95%

Supplier: Ambeed
Warning

A130740-10G A130740-1G A130740-250MG A130740-25G A130740-5G
77190-048EA 67 USD
77190-048 77190-050 77190-052 77190-054 77190-056
N,N'-Di-Boc-guanidine 95%
N,N'-Di-Boc-guanidine

Precaución: for Research Use Only

Formula: C₁₁H₂₁N₃O₄
MW: 259.30 g/mol
MDL Number: MFCD01863604
CAS Number: 154476-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR