6-Methoxy-1H-indazole-3-carbonitrile 98%

Supplier: Ambeed
Warning

A775709-1G
77171-214EA 229.69 USD
77171-214
6-Methoxy-1H-indazole-3-carbonitrile 98%
6-Methoxy-1H-indazole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₇N₃O
MW: 173.171 g/mol
MDL Number: MFCD06659784
CAS Number: 691900-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR