Zinc(II) Bis(trifluoromethanesulfonyl)imide 98%

Supplier: Ambeed
Danger

Synonyms: Zinc bis(trifluoromethylsulphonyl)imide, Bis(trifluoromethanesulphonyl)imide Zinc(II) Salt, Zinc(II) Triflimide

A311037-1G A311037-250MG A311037-25G A311037-5G
77167-108EA 12 USD
77167-108 77167-110 77167-112 77167-114
Zinc(II) Bis(trifluoromethanesulfonyl)imide 98%
Zinc(II) Bis(trifluoromethanesulfonyl)imide

Precaución: for Research Use Only

Formula: C₄F₁₂N₂O₈S₄Zn
MW: 625.65 g/mol
MDL Number: MFCD16621474
CAS Number: 168106-25-0
UN: 1759
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR