Fmoc-Cys(tert-butoxycarnylpropyl)-OH 95%

Supplier: Ambeed
Warning

A527507-100MG
77156-972EA 32 USD
77156-972
Fmoc-Cys(tert-butoxycarnylpropyl)-OH 95%
Fmoc-Cys(tert-butoxycarnylpropyl)-OH

Precaución: for Research Use Only

Formula: C₂₆H₃₁NO₆S
MW: 485.6 g/mol
MDL Number: MFCD11226818
CAS Number: 102971-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR