2,2'-Dibenzothiazole disulfide 96%

Supplier: Ambeed
Danger

Synonyms: 2,2'-Dithiobisbenzothiazole, 2,2-Dithiobis(benzothiazole), 2,2'-Dibenzothiazolyl disulphide

A475157-25G
77156-372EA 10 USD
77156-372
2,2'-Dibenzothiazole disulfide 96%
2,2'-Dibenzothiazole disulphide

Precaución: for Research Use Only

Formula: C₁₄H₈N₂S₄
MW: 332.4867 g/mol
MDL Number: MFCD00022874
CAS Number: 120-78-5
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR