Tetrahydro-1H-pyrrolizine-7a(5H)-carboxylic acid hydrochloride 95+%

Warning

Synonyms: Hexahydro-1H-pyrrolizine-7a-carboxylic acid hydrochloride

A379128-100MG
77155-284EA 151.41 USD
77155-284
Tetrahydro-1H-pyrrolizine-7a(5H)-carboxylic acid hydrochloride 95+%
Tetrahydro-1H-pyrrolizine-7a(5H)-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₄ClNO₂
MW: 191.66 g/mol
MDL Number: MFCD11035722
CAS Number: 165456-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR