Methyl 2-(aminomethyl)nicotinate hydrochloride 97%

Supplier: Ambeed
Warning

A105937-100MG A105937-250MG A105937-1G
103796-974EA 87 USD
103796-974 103822-722 77148-468 77148-470 77309-584
Methyl 2-(aminomethyl)nicotinate hydrochloride 97%
Methyl 2-(aminomethyl)nicotinate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁ClN₂O₂
MW: 202.64 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18803301
CAS Number: 151509-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR