RightCare No-Touch Infrared Forehead Thermometers
Supplier: OUTSOURCE PHARMACY SOLUTIONS, INC.
12894-005-3 12894-005-35CT
77148-070EA 30.6 USD
77148-070 77148-072
RightCare No-Touch Infrared Forehead Thermometers
Thermometers Digital Thermometers
Every family, office, business, and school needs a safe, accurate, and reliable thermometer. The RightCare digital non-contact Infrared Thermometer for adults and children is designed using the latest infrared technology. It includes the newest generation of core chip sensing that provides accurate readings within ±0.3 °C in under 0.5 seconds, without skin contact.
  • Non-contact sensor allows for safe, non-invasive body temperature readings at least 2" from forehead
  • Instant accurate readings in Fahrenheit or Celsius display on LCD screen within 0.5 seconds
More Product Information