(R)-3,3'-Dimethyl-[1,1'-binaphthalene]-2,2'-diol 98%

Supplier: Ambeed
Warning

A861799-100MG A861799-1G A861799-250MG A861799-5G
77146-952EA 27.81 USD
77146-952 77146-954 77146-956 77146-958
(R)-3,3'-Dimethyl-[1,1'-binaphthalene]-2,2'-diol 98%
(R)-3,3'-Dimethyl-[1,1'-binaphthalene]-2,2'-diol

Precaución: for Research Use Only

Formula: C₂₂H₁₈O₂
MW: 314.38 g/mol
MDL Number: MFCD00187966
CAS Number: 55515-98-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR