(R)-(+)-Chlocyphos 97%

Supplier: Ambeed
Warning

Synonyms: (4R)-4-(2-Chlorophenyl)-2-hydroxy-5,5-dimethyl-1,3,2-dioxaphosphinane 2-oxide, (4R)-(+)-4-(2-Chlorophenyl)-5,5-dimethyl-2-hydroxy-1,3,2-dioxaphosphinane 2-oxide

A474921-1G
77143-740EA 65.92 USD
77143-740
(R)-(+)-Chlocyphos 97%
(R)-(+)-Chlocyphos

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClO₄P
MW: 276.66 g/mol
MDL Number: MFCD00069409
CAS Number: 98674-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR