Anti-CSF2 Rat Monoclonal Antibody [clone: [BVD2-23B6]]

Supplier: Prosci
99-639
77120-684EA 445 USD
77120-684
Anti-CSF2 Rat Monoclonal Antibody [clone: [BVD2-23B6]]
Antibodies
Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: [BVD2-23B6]
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Rhesus Monkey, Cynomolgus Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR