1-Nitro-3-(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Warning

A186886-5G A186886-10G
103846-558EA 6 USD
103846-558 77084-250 77111-606
1-Nitro-3-(trifluoromethoxy)benzene 98%
1-Nitro-3-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₄F₃NO₃
MW: 207.11
Boiling Pt: 209.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00041011
CAS Number: 2995-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR