L(+)-Histidine 98%

Supplier: Ambeed
Warning

Synonyms: L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine

A170737-25G A170737-1000G A170737-100G A170737-10G A170737-500G A170737-5G
103844-810EA 5 USD
103844-810 77081-926 77110-976 77110-978 77110-980 77110-982 77301-070
L(+)-Histidine 98%
L(+)-Histidine

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Boiling Pt: 458.9 °C (760 mm Hg)
Melting Pt: 282 °C
Flash Pt: 282 °C
Storage Temperature: Ambient
MDL Number: MFCD00064315
CAS Number: 71-00-1
Merck Index: 13,04741

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR