Alendronate sodium trihydrate 98%

Supplier: Ambeed
Warning

Synonyms: (4-Amino-1-hydroxy-1-phosphonobutyl)phosphonic Acid Monosodium Salt, Sodium hydrogen (4-amino-1-hydroxy-1-phosphonobutyl)phosphonate trihydrate, Alendronic Acid Monosodium Salt, Alendronate sodium hydrate, Monosodium (4-Amino-1-hydroxy-1-phosphonobutyl)phosphonate

A136794-10G A136794-25G A136794-5G A136794-100G A136794-1000G A136794-500G
77109-588EA 10.3 USD
77109-588 77109-590 77109-592 77615-086 77685-180 77685-181
Alendronate sodium trihydrate 98%
Alendronate sodium trihydrate

Precaución: for Research Use Only

Formula: C₄H₁₂NaNO₇P₂·3H₂O
MW: 325.12 g/mol
MDL Number: MFCD01748233
CAS Number: 121268-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR