3-Benzyloxycyclobutanecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(benzyloxy)cyclobutane-1-carboxylic acid, 3-(Benzyloxy)cyclobutanecarboxylic acid

A128458-100G A128458-10G A128458-1G A128458-25G A128458-5G
77109-042EA 1213.34 USD
77109-042 77109-044 77109-046 77109-048 77109-050
3-Benzyloxycyclobutanecarboxylic acid 97%
3-Benzyloxycyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
MDL Number: MFCD00512751
CAS Number: 4958-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR