2,6-Dichlorobenzoyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 2,6-Dichlorobenzoyl Chloride

A738851-100G A738851-10G A738851-500G A738851-5G A738851-25G
77105-156EA 83 USD
77105-156 77105-158 77105-160 77105-162 77569-394
2,6-Dichlorobenzoyl chloride 98%
2,6-Dichlorobenzoyl chloride

Precaución: For research use only.

Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 260
Melting Pt: 18
MDL Number: MFCD00000662
CAS Number: 4659-45-4
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR