7-Fluoro-2,3-dihydropyrrolo[2,1-b]quinazolin-9(1H)-one 95%

Supplier: Ambeed
Warning

A608276-1G
77104-120EA 252.5 USD
77104-120
7-Fluoro-2,3-dihydropyrrolo[2,1-b]quinazolin-9(1H)-one 95%
7-Fluoro-2,3-dihydropyrrolo[2,1-b]quinazolin-9(1H)-one

Precaución: for Research Use Only

Formula: C₁₁H₉FN₂O
MW: 204.20 g/mol
MDL Number: MFCD02333781
CAS Number: 380638-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR