6-Bromo-1-methylindoline-2,3-dione 95%

Supplier: Ambeed
Warning

A271651-250MG
103813-052EA 63 USD
103813-052 77099-000
6-Bromo-1-methylindoline-2,3-dione 95%
6-Bromo-1-methylindoline-2,3-dione

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MW: 240.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11148035
CAS Number: 667463-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR