Ethyl-1-methylpipecolate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-Methyl-2-piperidinecarboxylate, 1-Methylpipecolic Acid Ethyl Ester, Ethyl 1-Methylpipecolate, Ethyl 1-Methylpipecolinate, 1-Methylpipecolinic Acid Ethyl Ester, 1-Methyl-2-piperidinecarboxylic Acid Ethyl Ester

A126931-5G
77094-812EA 13 USD
77094-812
Ethyl-1-methylpipecolate 98%
Ethyl-1-methylpipecolate

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
MDL Number: MFCD00006492
CAS Number: 30727-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR