Anti-Urokinase type Plasminogen Activator Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A101568-100
77072-900EA 484.2 USD
77072-900
Anti-Urokinase type Plasminogen Activator Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to uPA for IHC and ELISA with samples derived from Human.

  • Validated Applications: IHC, ELISA
  • Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000

Type: Primary
Antigen: Urokinase type Plasminogen Activator
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR