Nexus™ Horizontal Clean Bench, 6' Wide
Supplier: Labconco
5360210 5360300 5360310 5360500 5360510 5360600 5360610 5360700 5360710 5360800 5360000 5360010 5360100 5360110 5360200 5360810 5361000 5361010 5361100 5361110 5361200 5361210 5361300 5361310 5361500 5361510 5361600 5361610 5361700 5361710 5361800 5361810
77037-938EA 14980 USD
77037-938 77037-940 77037-942 77037-944 77037-946 77037-948 77037-950 77037-952 77037-954 77037-956 77037-928 77037-930 77037-932 77037-934 77037-936 77037-958 77037-960 77037-962 77037-964 77037-966 77037-968 77037-970 77037-972 77037-974 77037-976 77037-978 77037-980 77037-982 77037-984 77037-986 77037-988 77037-990
Nexus™ Horizontal Clean Bench, 6' Wide
Fume Hoods and Enclosures Laminar Flow Cabinets
Protects your non-hazardous applications with horizontal unidirectional (laminar) ISO Class 4 air across the work area.
  • Efficient unidirectional laminar airflow provided through CAP™ technology
  • Intelligent 5" touchscreen interface features intuitive monitoring capabilities
  • Reliable with long-life HEPA filters and maximum cleanability that stands up to disinfectants
The Nexus Horizontal clean bench is a laminar flow workstation designed to protect non-hazardous samples and processes from contamination.
The Nexus is the most advanced clean bench onMore Product Information