Nexus™ Horizontal Clean Bench, 5' Wide

Supplier: Labconco
5350000 5350010 5350100 5350110 5350200 5350210 5350300 5351610 5351700 5351710 5351800 5351810 5350310 5350500 5350510 5350600 5350610 5350700 5350710 5350800 5350810 5351000 5351010 5351100 5351110 5351200 5351210 5351300 5351310 5351500 5351510 5351600
77037-864EA 11670 USD
77037-864 77037-866 77037-868 77037-870 77037-872 77037-874 77037-876 77037-918 77037-920 77037-922 77037-924 77037-926 77037-878 77037-880 77037-882 77037-884 77037-886 77037-888 77037-890 77037-892 77037-894 77037-896 77037-898 77037-900 77037-902 77037-904 77037-906 77037-908 77037-910 77037-912 77037-914 77037-916
Nexus™ Horizontal Clean Bench, 5' Wide
Fume Hoods and Enclosures Laminar Flow Cabinets
Protects your non-hazardous applications with horizontal unidirectional (laminar) ISO Class 4 air across the work area.

  • Efficient unidirectional laminar airflow provided through CAP™ technology
  • Intelligent 5" touchscreen interface features intuitive monitoring capabilities
  • Reliable with long-life HEPA filters and maximum cleanability that stands up to disinfectants

The Nexus Horizontal clean bench is a laminar flow workstation designed to protect non-hazardous samples and processes from contamination. The Nexus provides an ISO 4 clean air environment.
More Product Information
Order Now

SPECIFICATIONS

Model 21" deep work area 26" deep work area
Environmentally Preferable
Dimensions 63.4×33.2×50.1" 63.4×33.2×50.1"

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR