4-(2-Hydroxyethyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4-(2-Hydroxyethyl)phenyl)boronic acid

A208452-100MG A208452-1G A208452-250MG A208452-5G
77030-412EA 19 USD
77030-412 77030-414 77030-416 77667-750
4-(2-Hydroxyethyl)phenylboronic acid 97%
4-(2-Hydroxyethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MW: 165.982 g/mol
MDL Number: MFCD03095135
CAS Number: 137756-89-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR