5-(3-Fluorophenyl)picolinic acid 95%

Supplier: Ambeed
Warning

A171777-250MG
77029-056EA 72 USD
77029-056
5-(3-Fluorophenyl)picolinic acid 95%
5-(3-Fluorophenyl)picolinic acid

Precaución: for Research Use Only

Formula: C₁₂H₈FNO₂
MW: 217.2 g/mol
MDL Number: MFCD12406462
CAS Number: 1158763-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR