2-Bromo-5,6-dihydrothieno[2,3-c]pyridin-7(4H)-one 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-7-oxo-4,5,6,7-tetrahydrothieno[2,3-c]pyridine

A154540-250MG
77028-306EA 96 USD
77028-306
2-Bromo-5,6-dihydrothieno[2,3-c]pyridin-7(4H)-one 95%
2-Bromo-5,6-dihydrothieno[2,3-c]pyridin-7(4H)-one

Precaución: for Research Use Only

Formula: C₇H₆BrNOS
MW: 232.098 g/mol
MDL Number: MFCD14584992
CAS Number: 960289-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR