2-(Boc-amino)-2-(2-naphthyl)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-((tert-Butoxycarbonyl)amino)-2-(naphthalen-2-yl)acetic acid

A919505-100MG A919505-1G A919505-250MG
77019-960EA 21 USD
77019-960 77019-962 77019-964
2-(Boc-amino)-2-(2-naphthyl)acetic acid 95%
2-(Boc-amino)-2-(2-naphthyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₉NO₄
MW: 301.337 g/mol
MDL Number: MFCD02683164
CAS Number: 33741-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR