3-(2-Hydroxyethylcarbamoyl)phenylboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: N-(2-Hydroxyethyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

A625441-100MG A625441-1G A625441-250MG A625441-5G
77018-140EA 37 USD
77018-140 77018-142 77018-144 77018-146
3-(2-Hydroxyethylcarbamoyl)phenylboronic acid pinacol ester 96%
3-(2-Hydroxyethylcarbamoyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₂BNO₄
MW: 291.1500 g/mol
MDL Number: MFCD06795647
CAS Number: 943911-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR