(1-Methylpiperidin-3-yl)methanamine 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Aminomethyl)-1-methylpiperidine

A454342-100MG A454342-1G A454342-250MG A454342-5G
77016-784EA 27 USD
77016-784 77016-786 77016-788 77016-790
(1-Methylpiperidin-3-yl)methanamine 95%
(1-Methylpiperidin-3-yl)methanamine

Precaución: for Research Use Only

Formula: C₇H₁₆N₂
MW: 128.2153 g/mol
MDL Number: MFCD06738902
CAS Number: 14613-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR