Methyl-2-chloro-4-(1-pyrrolidinyl)benzenecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-Chloro-4-(1-pyrrolidinyl)benzoate, Methyl 2-chloro-4-(pyrrolidin-1-yl)benzoate

A370130-100MG A370130-1G A370130-250MG
77015-968EA 12 USD
77015-968 77015-970 77015-972
Methyl-2-chloro-4-(1-pyrrolidinyl)benzenecarboxylate 97%
Methyl-2-chloro-4-(1-pyrrolidinyl)benzenecarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₄ClNO₂
MW: 239.7 g/mol
MDL Number: MFCD03844850
CAS Number: 175153-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR