Furo[3,2-b]pyridine-6-carboxylic acid 95%

Supplier: Ambeed
Warning

A262437-250MG
77014-212EA 78 USD
77014-212
Furo[3,2-b]pyridine-6-carboxylic acid 95%
Furo[3,2-b]pyridine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅NO₃
MDL Number: MFCD13193320
CAS Number: 122535-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR