3-(Difluoromethoxy)pyridin-2-amine 95%

Supplier: Ambeed
Warning

A251188-250MG
103811-970EA 66 USD
103811-970 103851-812 77014-002
3-(Difluoromethoxy)pyridin-2-amine 95%
3-(Difluoromethoxy)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₆F₂N₂O
MW: 160.121 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12405788
CAS Number: 947249-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR