7-(Trifluoromethoxy)isatin 98%

Supplier: Ambeed
Warning

Synonyms: 7-Trifluoromethoxyisatin

A142910-250MG
77010-456EA 11.67 USD
77010-456
7-(Trifluoromethoxy)isatin 98%
7-(Trifluoromethoxy)isatin

Precaución: for Research Use Only

Formula: C₉H₄F₃NO₃
MW: 231.1282 g/mol
MDL Number: MFCD06804535
CAS Number: 149125-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR