2,4,7-Trichloro-5H-pyrrolo[3,2-d]pyrimidine 95%

Supplier: Ambeed
Warning

A121248-250MG
77009-440EA 49 USD
77009-440
2,4,7-Trichloro-5H-pyrrolo[3,2-d]pyrimidine 95%
2,4,7-Trichloro-5H-pyrrolo[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₂Cl₃N₃
MW: 222.45 g/mol
MDL Number: MFCD22398070
CAS Number: 1923177-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR