2-Iodo-N-phenylacetamide 95%

Supplier: Ambeed
Warning

A109839-250MG
77008-862EA 90 USD
77008-862
2-Iodo-N-phenylacetamide 95%
2-Iodo-N-phenylacetamide

Precaución: for Research Use Only

Formula: C₈H₈INO
MW: 261.05 g/mol
MDL Number: MFCD16170501
CAS Number: 7212-28-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR