Swine Anti-Porcine Parvovirus Antibody IgG Titer Serologic Assay Kit (VP2)

Supplier: ReVacc Scientific

K10H97
77007-430EA 403.14 USD
77007-430
Swine Anti-Porcine Parvovirus Antibody IgG Titer Serologic Assay Kit (VP2)
Assays Immunological Assays

This kit is developed for serologic test for swine IgG titer of anti-PPV structural protein VP2 in serum/plasma or purified swine antibody (monoclonal and polyclonal) in vitro.


  • This kit was tested to bind to convalescent swine sera (n = 3) with sensitivity of 80%, specificity of 92% and Cohen's kappa of 0.85 for PPV VP2 protein


Assay principles: This kit is developed for a standard indirect-ELISA format, providing a rapid detection of anti-Porcine parvovirus (PPV) IgG in serum. The kit consists of high-bind detachable 96-well plate pre-coated with indicated protein, Positive control, negative control, an HRP-Anti-Swine IgG secondary antibody, TMB and dilution/wash/stop buffer.


Use protocol (4 simple steps):
1. Detach wells from plate based on your samples and keep at room temperature. Dilute your samples, positive and negative controls in sample dilution buffer and add them to wells (0.1 ml/well). Incubate at 37 °C for 1 hour or room temperature for 1.5 hour. Wash wells 4 times by wash solution (0.3 ml).
2. Dilute the secondary antibody HRP-Anti-Swine by wash solution and add to each well (0.1 ml). Incubate at room temperature for 0.5 to 1 hour. Avoid light by foil. Wash wells as mentioned.
3. Add TMB to each well (0.1 ml) and incubate at room temperature for 5 to 30 minutes.
4. Stop the substrate reaction by adding stop solution (0.1 ml/well). Absorbance (OD) is calculated as the absorbance at 450 nm. The OD value reflects the amount of antibody bound.


Storage and shipping condition: Shipment with room temperature. The unopened kit is stable for 6 months from the date of manufacture when stored at 2 to 8 °C. Avoid light. The shelf life is 60 days from the date of opening.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR