4-Hydroxy-o-anisidine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-3-methoxyphenol

A351304-100MG
77003-254EA 39 USD
77003-254
4-Hydroxy-o-anisidine 97%
4-Hydroxy-o-anisidine

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MW: 139.1519 g/mol
MDL Number: MFCD00272194
CAS Number: 61638-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR